Assay Details
Gene Symbol: Cblb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033238.1 | NP_001028410.1 | 5 - 6 | 872 | 128 | |
GenBank mRNA | AK147367.1 | - | 5 - 6 | 857 | 128 | |
AK156295.1 | - | 6 - 7 | 903 | 128 | ||
AK161486.1 | - | 3 - 4 | 458 | 128 | ||
AK170640.1 | - | 5 - 6 | 816 | 128 | ||
AK171350.1 | - | 5 - 6 | 817 | 128 | ||
BC150934.1 | - | 5 - 6 | 828 | 9087845 | 128 | |
BC150938.1 | - | 5 - 6 | 828 | 9087849 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
