Assay Details
Gene Symbol: Mast3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199308.2 | NP_955012.2 | 20 - 21 | 2632 | 92 | |
GenBank mRNA | BC024265.1 | - | 2 - 3 | 280 | 92 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC024265.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
