Assay Details
Gene Symbol: Nrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173440.2 | NP_775616.1 | 3 - 4 | 372 | 81 | |
GenBank mRNA | AK036273.1 | - | 3 - 4 | 373 | 81 | |
AK156233.1 | - | 3 - 4 | 351 | 81 | ||
AK157467.1 | - | 3 - 4 | 351 | 81 | ||
BC060232.1 | - | 2 - 3 | 170 | 6852643 | 81 | |
BC075638.1 | - | 3 - 4 | 249 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK036273.1, AK156233.1, BC060232.1, NM_173440.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
