Assay Details
Gene Symbol: Sacm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030692.4 | NP_109617.1 | 13 - 14 | 1102 | 148 | |
GenBank mRNA | AJ245720.1 | - | 13 - 14 | 1232 | 148 | |
AJ297526.1 | - | 13 - 14 | 1103 | 148 | ||
AK089174.1 | - | 13 - 14 | 1186 | 148 | ||
AK122390.1 | - | 13 - 14 | 1167 | 148 | ||
AK154759.1 | - | 13 - 14 | 1202 | 148 | ||
AK161111.1 | - | 13 - 14 | 1136 | 148 | ||
AK171080.1 | - | 13 - 14 | 1186 | 148 | ||
BC005489.1 | - | 4 - 5 | 311 | 148 | ||
BC117752.1 | - | 13 - 14 | 1103 | 40090556 | 148 | |
BC117753.1 | - | 13 - 14 | 1103 | 40090558 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
