Assay Details
Gene Symbol: Sidt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159419.1 | NP_001152891.1 | 23 - 24 | 2521 | 67 | |
NM_198034.3 | NP_932151.2 | 22 - 23 | 2506 | 67 | ||
GenBank mRNA | AK034344.1 | - | 8 - 9 | 671 | 67 | |
AK046836.1 | - | 23 - 24 | 2881 | 67 | ||
AK171920.1 | - | 22 - 23 | 2659 | 67 | ||
AY562213.1 | - | 12 - 13 | 1065 | 67 | ||
AY562214.1 | - | 12 - 13 | 1065 | 67 | ||
BC079584.1 | - | 22 - 23 | 2506 | 5704672 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
