Assay Details
Gene Symbol: Dzip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110017.1 | NP_001103487.1 | 26 - 27 | 3286 | 94 | |
NM_027341.2 | NP_081617.1 | 25 - 26 | 2668 | 94 | ||
GenBank mRNA | AK122344.1 | - | 3 - 4 | 1829 | 94 | |
BC117953.1 | - | 25 - 26 | 2491 | 40098141 | 94 | |
BC117954.1 | - | 25 - 26 | 2497 | 40098143 | 94 | |
EU233997.1 | - | 18 - 19 | 2488 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
