Assay Details
Gene Symbol: Tnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110147.1 | NP_001103617.1 | 2 - 3 | 647 | 120 | |
NM_016788.3 | NP_058068.2 | 2 - 3 | 647 | 120 | ||
GenBank mRNA | AF037260.1 | - | 2 - 3 | 120 | ||
BC052421.1 | - | 2 - 3 | 647 | 6400209 | 120 | |
DQ666696.1 | - | 2 - 3 | 458 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
