Assay Details
Gene Symbol: Adgrg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172825.3 | NP_766413.2 | 4 - 5 | 643 | 148 | |
GenBank mRNA | AK033593.1 | - | 4 - 5 | 644 | 148 | |
BC132302.1 | - | 4 - 5 | 468 | 40130579 | 148 | |
BC137963.1 | - | 4 - 5 | 468 | 8860982 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
