Assay Details
Gene Symbol: Uso1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019490.1 | NP_062363.1 | 8 - 9 | 830 | 157 | |
GenBank mRNA | AK144638.1 | - | 8 - 9 | 821 | 157 | |
AK156070.1 | - | 8 - 9 | 823 | 157 | ||
AK166587.1 | - | 9 - 10 | 790 | 157 | ||
AK172425.1 | - | 8 - 9 | 821 | 157 | ||
BC016069.1 | - | 8 - 9 | 830 | 4480992 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
