Assay Details
Gene Symbol: Cxcl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF227743.1 | - | 3 - 4 | 317 | 144 | |
AJ243095.1 | - | 2 - 3 | 168 | 144 | ||
AK146144.1 | - | 3 - 4 | 354 | 144 | ||
M86829.1 | - | 3 - 4 | 346 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
