Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
TC0400000644.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037127.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037127.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037128.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037129.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037129.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001037130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001165996.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_001165996.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_010944.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004558.mm.1 |
NM_010944.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
TC0400000644.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037127.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037127.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037128.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037129.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037129.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001037130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001165996.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_001165996.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_010944.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004554.mm.1 |
NM_010944.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
TC0400000644.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037127.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037127.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037128.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037129.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037129.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001037130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001165996.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_001165996.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_010944.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004555.mm.1 |
NM_010944.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
TC0400000644.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037127.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037127.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037128.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037129.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037129.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037130.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001037130.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001165996.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_001165996.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_010944.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400004557.mm.1 |
NM_010944.2 |
Refseq |