Assay Details
Gene Symbol: Agmat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081408.1 | NP_001074877.1 | 6 - 7 | 1140 | 66 | |
GenBank mRNA | AK017155.1 | - | 6 - 7 | 1037 | 66 | |
BC025916.1 | - | 3 - 4 | 357 | 66 | ||
BC037459.1 | - | 3 - 4 | 313 | 66 | ||
BC115693.2 | - | 6 - 7 | 1037 | 40104399 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | BC115693.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | NM_001081408.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | AK017155.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | BC025916.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | BC037459.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | BC115693.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | NM_001081408.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | AK017155.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | BC025916.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P400463 | BC037459.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 198836 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 272799 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270632 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612952 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854653 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111430 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587715 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914124 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50622 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | BC115693.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | NM_001081408.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | AK017155.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | BC025916.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | BC037459.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | BC115693.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | NM_001081408.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | AK017155.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | BC025916.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P400463 | BC037459.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05431358 | - | - |
Whole Mouse Genome G4122A | A_51_P400463 | BC115693.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | NM_001081408.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | AK017155.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | BC025916.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | BC037459.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | BC115693.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | NM_001081408.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | AK017155.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | BC025916.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P400463 | BC037459.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365709 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365728 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365716 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23365720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03416655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03416649 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04017151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03961895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03544325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365706 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03652081 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03490696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03490695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03652080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03311148 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03259791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03311154 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03311152 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03436235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03380596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03439394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03591934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03311144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04018665 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03624635 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03608412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03608418 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03380599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03652082 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03553736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365762 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03416654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03380594 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03403573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03743886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365780 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03403574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23365753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04090874 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10510028 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510030 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510026 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510032 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510031 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510027 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510029 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10510033 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427214_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10510025 | - | - |
Mouse Genome 430 2.0 Array | 1427214_at | - | - |
Mouse Genome 430 2.0 Array | 1427214_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75986_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75986_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400014942.mm.1 | TC0400001800.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400014942.mm.1 | NM_001081408.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400014942.mm.1 | NM_001081408.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400014943.mm.1 | TC0400001800.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400014943.mm.1 | NM_001081408.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400014943.mm.1 | NM_001081408.1 | Refseq |
Mouse Expression Set 430 | 1427214_at | - | - |
Mouse Expression Set 430 | 1427214_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427214_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427214_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427214_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427214_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 847544 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2642426 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2642426 | - | - |
