Assay Details
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037129.1 | NP_001032206.1 | 11 - 12 | 1544 | 59 | |
NM_001037130.1 | NP_001032207.1 | 10 - 11 | 1514 | 59 | ||
GenBank mRNA | AB691772.1 | - | 10 - 11 | 1364 | 59 | |
AY955298.1 | - | 11 - 12 | 1394 | 59 | ||
AY956404.1 | - | 10 - 11 | 1364 | 59 | ||
BC100342.1 | - | 11 - 12 | 1468 | 30935290 | 59 | |
U37708.1 | - | 10 - 11 | 1512 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
