Assay Details
Gene Symbol: Musk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037127.2 | NP_001032204.2 | 12 - 13 | 1607 | 78 | |
NM_001037128.1 | NP_001032205.1 | 12 - 13 | 1562 | 78 | ||
NM_010944.2 | NP_035074.2 | 11 - 12 | 1532 | 78 | ||
GenBank mRNA | AY955296.1 | - | 12 - 13 | 1457 | 78 | |
AY955297.1 | - | 12 - 13 | 1412 | 78 | ||
AY956403.1 | - | 11 - 12 | 1382 | 78 | ||
U37709.1 | - | 11 - 12 | 1530 | 78 | ||
X86444.1 | - | 10 - 11 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
