Assay Details
Gene Symbol: Ctnnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165902.1 | NP_001159374.1 | 4 - 5 | 582 | 72 | |
NM_007614.3 | NP_031640.1 | 4 - 5 | 782 | 72 | ||
GenBank mRNA | AK018515.1 | - | 4 - 5 | 696 | 72 | |
AK020013.1 | - | 4 - 5 | 697 | 72 | ||
AK035311.1 | - | 4 - 5 | 695 | 72 | ||
AK132398.1 | - | 4 - 5 | 763 | 72 | ||
AK133656.1 | - | 4 - 5 | 695 | 72 | ||
AK135743.1 | - | 1 - 2 | 249 | 72 | ||
AY751549.1 | - | 4 - 5 | 707 | 72 | ||
BC043078.1 | - | 1 - 2 | 184 | 72 | ||
BC048153.1 | - | 4 - 5 | 707 | 5709247 | 72 | |
BC053065.1 | - | 4 - 5 | 707 | 5709247 | 72 | |
M90364.1 | - | 4 - 5 | 592 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
