Assay Details
Gene Symbol: Gm15713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046026.1 | 3 - 4 | 330 | 104 | ||
GenBank mRNA | AK042323.1 | - | 3 - 4 | 331 | 104 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger and BTB domain containing 20 | Zbtb20 |
1300017A20Rik, 7330412A13Rik, A930017C21Rik, D16Wsu73e, DPZF, HOF, ODA-8S, Oda8, Zfp288 |
Anti-sense | Chr. 16 - 42875853 - 43621944 on NCBI Build GRCm38 | |
Downstream | microRNA 568 | Mir568 |
Mirn568, mmu-mir-568 |
224.9kb | Anti-sense | Chr. 16 - 43640655 - 43640737 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
