Assay Details
Gene Symbol: Gm19619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040428.1 | 3 - 4 | 1464 | 56 | ||
GenBank mRNA | AK050091.1 | - | 3 - 4 | 1465 | 56 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | amphiregulin | Areg |
AR, Mcub, Sdgf |
-125.3kb | Anti-sense | Chr. 5 - 91139615 - 91148432 on NCBI Build GRCm38 |
Downstream |
betacellulin, epidermal growth factor family member |
Btc | Bcn | 76.4kb | Sense | Chr. 5 - 91357261 - 91402994 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400000269.rn.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589979 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 305296 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 927162 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 80011 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605850 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 761261 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123686 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110275 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109644 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05653311 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109644 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125726 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109644 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04912088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04428092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04123092 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04878625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887897 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887857 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887890 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887915 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887790 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887845 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04878623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04878633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887854 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04878624 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04852141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04608057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04123093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04630893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04102627 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887903 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04852146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887923 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887849 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887793 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887927 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04878636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23887847 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0500024124.mm.1 | TC0500002737.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500024124.mm.1 | NR_040428.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024124.mm.1 | NR_040428.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024122.mm.1 | TC0500002737.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500024122.mm.1 | NR_040428.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024122.mm.1 | NR_040428.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024123.mm.1 | TC0500002737.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500024123.mm.1 | NR_040428.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500024123.mm.1 | NR_040428.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 575579 | - | - |
