Assay Details
Gene Symbol: Slc44a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133891.3 | NP_598652.3 | 14 - 15 | 2130 | 75 | |
GenBank mRNA | AY249865.1 | - | 14 - 15 | 1886 | 75 | |
AY249866.1 | - | 14 - 15 | 1859 | 75 | ||
BC010258.1 | - | 2 - 3 | 324 | 75 | ||
BC025941.1 | - | 5 - 6 | 692 | 75 | ||
BC113167.1 | - | 14 - 15 | 1886 | 40059854 | 75 | |
BC113169.1 | - | 14 - 15 | 1886 | 40059860 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
