Assay Details
Gene Symbol: Slc44a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159633.1 | NP_001153105.1 | 2 - 3 | 389 | 72 | |
NM_133891.3 | NP_598652.3 | 2 - 3 | 389 | 72 | ||
GenBank mRNA | AY249865.1 | - | 2 - 3 | 145 | 72 | |
AY249866.1 | - | 2 - 3 | 118 | 72 | ||
BC113167.1 | - | 2 - 3 | 145 | 40059854 | 72 | |
BC113169.1 | - | 2 - 3 | 145 | 40059860 | 72 | |
CJ054525.1 | - | 2 - 3 | 349 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
