Assay Details
Gene Symbol: 9530059O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK049978.1 | - | 461 | 80 | ||
AK079255.1 | - | 546 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | microRNA 138-1 | Mir138-1 |
Mirn138, Mirn138-1, mir-138-1 |
9.7kb | Sense | Chr. 9 - 122682876 - 122682974 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
