Assay Details
Gene Symbol: Gm31410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076263.1 | - | 3 - 4 | 886 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 4668 | Gm4668 | -29.7kb | Sense | Chr. 9 - 116184530 - 116195049 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA D730003K21 gene | D730003K21Rik | 77.6kb | Sense | Chr. 9 - 116327308 - 116339659 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
