Assay Details
Gene Symbol: Gm33733 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK081111.1 | - | 1 - 2 | 1889 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nudix (nucleoside diphosphate linked moiety X)-type motif 10 | Nudt10 |
AI853080, Dipp3a |
-31.2kb | Anti-sense | Chr. X - 6168696 - 6173044 on NCBI Build GRCm38 |
Downstream | bone morphogenetic protein 15 | Bmp15 |
AU015375, AU018861, AU021453, Bmp-15, C86824, C87336, GDF-9B |
101.0kb | Anti-sense | Chr. X - 6314106 - 6320723 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
