Assay Details
Gene Symbol: Rad23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009011.4 | NP_033037.2 | 1 - 2 | 384 | 97 | |
GenBank mRNA | AK084917.1 | - | 1 - 2 | 376 | 97 | |
AK142280.1 | - | 1 - 2 | 376 | 97 | ||
AK150089.1 | - | 1 - 2 | 376 | 97 | ||
AK157252.1 | - | 1 - 2 | 346 | 97 | ||
AK160880.1 | - | 1 - 2 | 376 | 97 | ||
AK160890.1 | - | 1 - 2 | 362 | 97 | ||
AK160973.1 | - | 1 - 2 | 362 | 97 | ||
AK167582.1 | - | 1 - 2 | 372 | 97 | ||
BC027747.1 | - | 1 - 2 | 373 | 4459657 | 97 | |
BC068193.1 | - | 1 - 2 | 343 | 6813997 | 97 | |
X92411.1 | - | 1 - 2 | 64 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
