Assay Details
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173788.3 | NP_776149.1 | 5 - 6 | 1470 | 79 | |
GenBank mRNA | BC042470.1 | - | 5 - 6 | 1470 | 5137261 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
