Assay Details
Gene Symbol: Gba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172692.3 | NP_766280.2 | 4 - 5 | 1280 | 65 | |
GenBank mRNA | AK089192.1 | - | 4 - 5 | 966 | 65 | |
AK158174.1 | - | 4 - 5 | 1280 | 65 | ||
AK173213.1 | - | 4 - 5 | 1266 | 65 | ||
AK178058.1 | - | 1 - 2 | 196 | 65 | ||
AK214862.1 | - | 1 - 2 | 194 | 65 | ||
AK217543.1 | - | 1 - 2 | 196 | 65 | ||
BC031397.1 | - | 4 - 5 | 953 | 65 | ||
BC034105.1 | - | 4 - 5 | 505 | 65 | ||
BC056935.1 | - | 4 - 5 | 803 | 5703478 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
