Assay Details
Gene Symbol: Dsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023842.2 | NP_076331.2 | 9 - 10 | 1463 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
