Assay Details
Gene Symbol: Riok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024242.3 | NP_077204.2 | 11 - 12 | 1564 | 72 | |
GenBank mRNA | AK017312.1 | - | 11 - 12 | 1259 | 72 | |
AK152707.1 | - | 11 - 12 | 1313 | 72 | ||
AK161315.1 | - | 12 - 13 | 1807 | 72 | ||
BC002158.1 | - | 5 - 6 | 521 | 3485522 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
