Assay Details
Gene Symbol: Nup153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175749.2 | NP_786925.2 | 10 - 11 | 1638 | 136 | |
GenBank mRNA | BC052173.1 | - | 10 - 11 | 1376 | 6516328 | 136 |
CA326562.1 | - | 5 - 6 | 330 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
