Assay Details
Gene Symbol: Tert | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009354.1 | NP_033380.1 | 7 - 8 | 2394 | 59 | |
GenBank mRNA | AF051911.1 | - | 7 - 8 | 2394 | 59 | |
AF073311.1 | - | 7 - 8 | 2365 | 59 | ||
BC082327.1 | - | 8 - 9 | 1896 | 59 | ||
BC127068.1 | - | 8 - 9 | 1682 | 40110173 | 59 | |
FJ210936.1 | - | 3 - 4 | 390 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
