Assay Details
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009651.4 | NP_033781.2 | 1 - 2 | 145 | 70 | |
GenBank mRNA | BC130271.1 | - | 1 - 2 | 77 | 40143193 | 70 |
U10341.1 | - | 1 - 2 | 150 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC130271.1, NM_009651.4, U10341.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
