Assay Details
Gene Symbol: 4921525O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045061.1 | 4 - 5 | 1011 | 97 | ||
GenBank mRNA | AK014969.1 | - | 4 - 5 | 1012 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 26651 |
Gm26651 | -139.6kb | Sense | Chr. 13 - 51973214 - 51978536 on NCBI Build GRCm38 | |
Downstream |
DIRAS family, GTP-binding RAS-like 2 |
Diras2 |
2900052J15Rik, AI414999 |
384.8kb | Anti-sense | Chr. 13 - 52504375 - 52531242 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
