Assay Details
Gene Symbol: 4921525O09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045061.1 | 7 - 8 | 2153 | 66 | ||
GenBank mRNA | AK014969.1 | - | 7 - 8 | 2154 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 26651 |
Gm26651 | -143.6kb | Sense | Chr. 13 - 51973214 - 51978536 on NCBI Build GRCm38 | |
Downstream |
DIRAS family, GTP-binding RAS-like 2 |
Diras2 |
2900052J15Rik, AI414999 |
380.2kb | Anti-sense | Chr. 13 - 52504375 - 52531242 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2427175 | AK014969.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2427175 | AV256185.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191198 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748856 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810862 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586697 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 153662 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 317612 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601540 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165404 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 4193 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2427175 | AK014969.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2427175 | AV256185.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02691169 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02691194 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02691144 | - | - |
Whole Mouse Genome G4122A | A_52_P668206 | AK014969.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114713 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02691194 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02691144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036417 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10700673 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036265 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515527 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036438 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036307 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036467 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10536790 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10721089 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10793870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11059177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10700662 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10835362 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10850848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515518 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10536787 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11059174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036473 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036462 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10536789 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036283 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036469 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10536781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10835369 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10700660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036407 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515535 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10536779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10850847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11059203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10536776 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515525 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11059167 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036446 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036388 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036454 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036405 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036400 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10850854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10632712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036262 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036288 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10743434 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515532 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036474 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515524 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515519 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036442 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10787322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11059164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036481 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10536796 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10721092 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036390 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036453 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036422 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036471 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036432 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10515526 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10700659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10632713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036393 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10835363 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10850853 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10937543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036306 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11059189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036311 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036441 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036444 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11059175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P29036416 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1429472_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1447836_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1429472_at | - | - |
Mouse Genome 430 2.0 Array | 1447836_x_at | - | - |
Mouse Genome 430 2.0 Array | 1429472_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1447836_PM_x_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300003851.mm.1 | TC1300000609.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300003851.mm.1 | NR_045061.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300003851.mm.1 | NR_045061.1 | Refseq |
Mouse Expression Set 430 | 1429472_PM_at | - | - |
Mouse Expression Set 430 | 1447836_PM_x_at | - | - |
Mouse Expression Set 430 | 1447836_x_at | - | - |
Mouse Expression Set 430 | 1429472_at | - | - |
Murine Genome U74v2 Set | 166290_r_at | - | - |
Murine Genome U74v2 Set | 166290_PM_r_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429472_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447836_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429472_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447836_PM_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 794783 | - | - |
