Assay Details
Gene Symbol: Secisbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK008918.1 | - | 1 - 2 | 165 | 114 | |
AK030021.1 | - | 10 - 11 | 1459 | 114 | ||
AK156076.1 | - | 10 - 11 | 1535 | 114 | ||
AK168634.1 | - | 10 - 11 | 1492 | 114 | ||
BC125391.1 | - | 10 - 11 | 1515 | 40129906 | 114 | |
BC125393.1 | - | 10 - 11 | 1512 | 40129908 | 114 | |
BC144795.1 | - | 10 - 11 | 1510 | 114 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
