Assay Details
Gene Symbol: Eda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177937.1 | NP_001171408.1 | 2 - 3 | 676 | 78 | |
NM_001177938.1 | NP_001171409.1 | 2 - 3 | 676 | 78 | ||
NM_001177939.1 | NP_001171410.1 | 2 - 3 | 676 | 78 | ||
NM_001177942.1 | NP_001171413.1 | 2 - 3 | 676 | 78 | ||
NM_001177943.1 | NP_001171414.1 | 2 - 3 | 676 | 78 | ||
NM_001177944.1 | NP_001171415.1 | 2 - 3 | 676 | 78 | ||
NM_010099.2 | NP_034229.1 | 2 - 3 | 676 | 78 | ||
GenBank mRNA | AF004435.1 | - | 2 - 3 | 642 | 78 | |
AF016628.1 | - | 2 - 3 | 676 | 78 | ||
AF016629.1 | - | 2 - 3 | 676 | 78 | ||
AJ243657.1 | - | 2 - 3 | 663 | 78 | ||
AJ243658.1 | - | 2 - 3 | 624 | 78 | ||
AK137551.1 | - | 2 - 3 | 585 | 78 | ||
BC125387.1 | - | 2 - 3 | 567 | 40129902 | 78 | |
BC144790.1 | - | 2 - 3 | 567 | 9053338 | 78 | |
BC144791.1 | - | 2 - 3 | 567 | 9053339 | 78 | |
DQ168876.1 | - | 2 - 3 | 676 | 78 | ||
DQ168877.1 | - | 2 - 3 | 676 | 78 | ||
DQ168880.1 | - | 2 - 3 | 676 | 78 | ||
DQ168881.1 | - | 2 - 3 | 676 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
