Assay Details
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 7 - 8 | 1393 | 60 | |
NM_019867.2 | NP_063920.2 | 5 - 6 | 1030 | 60 | ||
GenBank mRNA | AJ238898.1 | - | 5 - 6 | 1011 | 60 | |
AK138433.1 | - | 5 - 6 | 999 | 60 | ||
AY038025.1 | - | 5 - 6 | 1005 | 60 | ||
BC039279.1 | - | 7 - 8 | 1393 | 4973826 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
