Assay Details
Gene Symbol: Senp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027838.3 | NP_082114.1 | 1 - 2 | 74 | 63 | |
GenBank mRNA | AK029856.1 | - | 1 - 2 | 64 | 63 | |
BC037443.1 | - | 1 - 2 | 46 | 3599077 | 63 | |
DV645349.1 | - | 1 - 2 | 74 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC037443.1, NM_027838.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
