Assay Details
Gene Symbol: C78653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043315.1 | - | 1 - 2 | 709 | 117 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 38403 |
Gm38403 | C230029M16 | -91.2kb | Sense | Chr. 10 - 118920381 - 118927143 on NCBI Build GRCm38 |
Downstream |
predicted gene, 33677 |
Gm33677 | 6.6kb | Sense | Chr. 10 - 119028287 - 119053655 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
