Assay Details
Gene Symbol: Opn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK027928.1 | - | 1 - 2 | 90 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LIM domain binding 3 | Ldb3 |
AW742271, PDLIM6, ZASP |
-1.9kb | Sense | Chr. 14 - 34526699 - 34588698 on NCBI Build GRCm38 |
Downstream | wings apart-like homolog (Drosophila) | Wapal |
A530089A20Rik, BC037674, DIF-2, FOE, WAPL |
79.6kb | Anti-sense | Chr. 14 - 34671487 - 34747983 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
