Assay Details
Gene Symbol: Macrod2os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK031201.1 | - | 5 - 6 | 1723 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MACRO domain containing 2 | Macrod2 |
1110033L15Rik, 2610107G07Rik, 2900006F19Rik |
Anti-sense | Chr. 2 - 140395356 - 142392966 on NCBI Build GRCm38 | |
Downstream | kinesin family member 16B | Kif16b |
8430434E15Rik, AA623607, AI789011, C80253, C80902, mKIAA1590 |
1.4mb | Sense | Chr. 2 - 142618334 - 142901595 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
