Assay Details
Gene Symbol: 4933424M12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016731.1 | - | 1 - 2 | 1120 | 88 | |
AK016899.1 | - | 1 - 2 | 1161 | 88 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
DMRT-like family B with proline-rich C-terminal, 1 |
Dmrtb1 |
Dmrt6, Prp13 |
-103.6kb | Anti-sense | Chr. 4 - 107676288 - 107684243 on NCBI Build GRCm38 |
Downstream |
low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, opposite strand 3 |
Lrp8os3 |
Gm12908, OTTMUSG00000009011 |
3.2kb | Anti-sense | Chr. 4 - 107793156 - 107802171 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965512 | AK016731.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1965512 | AK016899.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965512 | AK016731.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1965512 | AK016899.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00955394 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04091229 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03912213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03884503 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03514560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03387227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03387226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03328247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03328245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03438774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03507354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03772342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03272933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03514562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192790 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03514558 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192755 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192777 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03514561 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192802 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03514568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192767 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03884506 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03438779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03520766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03584098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03884500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03520956 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23192745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03912211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03328248 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03347837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03604650 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03272935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03328249 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1430218_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1430218_at | - | - |
Mouse Genome 430 2.0 Array | 1430218_PM_at | - | - |
Mouse Expression Set 430 | 1430218_at | - | - |
Mouse Expression Set 430 | 1430218_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430218_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430218_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 854893 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 104590347 | - | - |
