Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
TC0900003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001040632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001040632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001190313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001190313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001190314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001190314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001190319.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_001190319.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_145613.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_145613.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_147126.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003310.mm.1 |
NM_147126.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
TC0900003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001040631.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001040631.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001040632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001040632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001190313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001190313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001190314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001190314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001190319.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_001190319.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_145613.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_145613.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_147126.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003308.mm.1 |
NM_147126.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
TC0900003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001040632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001040632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001190313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001190313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001190314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001190314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001190319.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_001190319.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_145613.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_145613.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_147126.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003309.mm.1 |
NM_147126.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
TC0900003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001040631.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001040631.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001040632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001040632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001190313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001190313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001190314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_001190314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_145613.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_145613.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_147126.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003306.mm.1 |
NM_147126.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
TC0900003323.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001040631.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001040631.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001040632.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001040632.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001190313.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001190313.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001190314.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001190314.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001190319.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_001190319.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_145613.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_145613.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_147126.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003313.mm.1 |
NM_147126.3 |
Refseq |