Assay Details
Gene Symbol: Kansl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289437.1 | NP_001276366.1 | 1 | 85 | 101 | |
NM_001289438.1 | NP_001276367.1 | 1 | 85 | 101 | ||
NM_001289439.1 | NP_001276368.1 | 1 | 85 | 101 | ||
NM_133714.5 | NP_598475.2 | 1 | 85 | 101 | ||
GenBank mRNA | AK046649.1 | - | 1 | 55 | 101 | |
AK089025.1 | - | 1 | 74 | 101 | ||
AK150748.1 | - | 1 | 80 | 101 | ||
AK151604.1 | - | 1 | 81 | 101 | ||
AK152604.1 | - | 1 | 80 | 101 | ||
AK153007.1 | - | 1 | 80 | 101 | ||
AK162714.1 | - | 1 | 59 | 101 | ||
AK167429.1 | - | 1 | 60 | 101 | ||
AK168499.1 | - | 1 | 64 | 101 | ||
BC003301.1 | - | 1 | 61 | 3584464 | 101 | |
BC125631.1 | - | 1 | 37 | 40130146 | 101 | |
BC125633.1 | - | 1 | 37 | 40130148 | 101 | |
BC145047.1 | - | 1 | 37 | 101 | ||
CJ059354.1 | - | 1 | 86 | 101 |
Gene Symbol: 2610028H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK160214.1 | - | 1 | 101 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK046649.1, AK167429.1, BC003301.1, BC125631.1, BC125633.1, NM_001289437.1, NM_001289438.1, NM_001289439.1, NM_133714.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
