Assay Details
Gene Symbol: Hrk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007545.2 | NP_031571.1 | 2 | 808 | 88 | |
GenBank mRNA | D83698.1 | - | 2 | 807 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025038 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025034 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025046 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025048 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025030 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025031 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025047 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025039 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025037 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025029 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025049 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025036 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2083559 | NM_007545.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533639 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917314 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764529 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780832 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 682061 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164315 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484879 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 238692 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72559 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855049 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 345920 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98754 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136782 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139203 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122503 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2083559 | NM_007545.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025051 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025025 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025038 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025022 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025045 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025035 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025041 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025028 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025044 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025046 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025047 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025039 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025013 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025032 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24025024 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122503 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01199697 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139203 | - | - |
Whole Mouse Genome G4122A | A_51_P509137 | NM_007545.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509137 | BC128022.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P509137 | D83698.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135127 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122503 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139203 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04833490 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04833484 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04833489 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025029 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025046 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025034 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025039 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025038 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025035 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025031 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025049 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025032 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025037 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025030 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025048 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025036 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24025047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24024984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24024986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24024990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025067 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24024988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24024996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24024998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24024992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24025106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04437924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025073 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025058 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025067 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24024997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04321841 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04184681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025082 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04141707 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04287493 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24024998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04102273 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04227473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04794602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04102278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04705568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04370526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04276455 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025077 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04141704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04437925 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04705569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04519177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04535219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P24025003 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10524970 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524972 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10524971 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1439854_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450366_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10524969 | - | - |
Mouse Genome 430 2.0 Array | 1439854_at | - | - |
Mouse Genome 430 2.0 Array | 1450366_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1450366_at | - | - |
Mouse Genome 430 2.0 Array | 1439854_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12123_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12123_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500011451.mm.1 | TC0500001307.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500011451.mm.1 | NM_007545.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500011451.mm.1 | NM_007545.2 | Refseq |
Mouse Expression Set 430 | 1450366_PM_at | - | - |
Mouse Expression Set 430 | 1450366_at | - | - |
Mouse Expression Set 430 | 1439854_at | - | - |
Mouse Expression Set 430 | 1439854_PM_at | - | - |
Murine Genome U74v2 Set | 101189_PM_at | - | - |
Murine Genome U74v2 Set | 101189_at | - | - |
Mouse Genome 430A 2.0 Array | 1450366_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1450366_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450366_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450366_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439854_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439854_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 476307 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2030021 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2030021 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002030021 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1229407 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1229407 | - | - |
mouse-6 v1.1 (Illumina) | scl27311.3.1_141-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2030021 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl27311.3.1_141-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2030021 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1229407 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1229407 | - | - |
