Assay Details
Gene Symbol: C1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183175.4 | NP_898998.2 | 2 - 3 | 226 | 74 | |
GenBank mRNA | BC115632.1 | - | 2 - 3 | 211 | 40102920 | 74 |
BC115633.1 | - | 2 - 3 | 211 | 40102924 | 74 | |
DQ002401.1 | - | 2 - 3 | 226 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
