Assay Details
Gene Symbol: Akap7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018747.4 | NP_061217.3 | 3 - 4 | 410 | 80 | |
GenBank mRNA | AY301068.1 | - | 3 - 4 | 417 | 80 | |
DV647989.1 | - | 3 - 4 | 489 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
