Assay Details
Gene Symbol: Vwde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013757.2 | NP_001013779.2 | 6 - 7 | 997 | 95 | |
GenBank mRNA | BC076621.1 | - | 6 - 7 | 997 | 95 | |
BC151200.1 | - | 6 - 7 | 906 | 9088088 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
