Assay Details
Gene Symbol: Slc47a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033542.2 | NP_001028714.1 | 5 - 6 | 612 | 89 | |
GenBank mRNA | AK133432.1 | - | 5 - 6 | 613 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
