Assay Details
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037326.2 | NP_001032403.1 | 2 | 292 | 85 | |
GenBank mRNA | AK083093.1 | - | 2 | 158 | 85 | |
AK164375.1 | - | 1 | 4358 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
