Assay Details
Gene Symbol: Copg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017477.2 | NP_059505.1 | 3 - 4 | 265 | 107 | |
NM_201244.1 | NP_957696.1 | 3 - 4 | 233 | 107 | ||
GenBank mRNA | AF187079.1 | - | 3 - 4 | 264 | 107 | |
AK018151.1 | - | 3 - 4 | 193 | 105 | ||
AK049417.1 | - | 3 - 4 | 216 | 107 | ||
AK077461.1 | - | 3 - 4 | 212 | 107 | ||
AK080451.1 | - | 3 - 4 | 237 | 107 | ||
AK147895.1 | - | 3 - 4 | 278 | 107 | ||
AK151284.1 | - | 3 - 4 | 270 | 107 | ||
AK151816.1 | - | 3 - 4 | 270 | 107 | ||
AK153142.1 | - | 3 - 4 | 270 | 107 | ||
AK164280.1 | - | 3 - 4 | 235 | 107 | ||
AK169256.1 | - | 3 - 4 | 310 | 107 | ||
AK171950.1 | - | 3 - 4 | 244 | 107 | ||
AK172519.1 | - | 3 - 4 | 281 | 107 | ||
BC024686.1 | - | 3 - 4 | 236 | 3992144 | 107 | |
BC024896.1 | - | 3 - 4 | 233 | 4189311 | 107 | |
BC056168.1 | - | 3 - 4 | 233 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
