Assay Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047159.2 | NP_001040624.1 | 1 - 2 | 290 | 86 | |
GenBank mRNA | AJ010045.1 | - | 1 - 2 | 253 | 86 | |
AK082128.1 | - | 1 - 2 | 287 | 86 | ||
AK088632.1 | - | 1 - 2 | 254 | 86 | ||
AK139921.1 | - | 1 - 2 | 254 | 86 | ||
BC004699.1 | - | 1 - 2 | 264 | 3499258 | 86 | |
CN702942.1 | - | 1 - 2 | 151 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
